EMAIL THIS PAGE TO A FRIEND
Z214086 Aldrich
Extractor Kit 2000
replacement filter
Purchase
Properties
Related Categories Extraction Kit, Fume Hood, Labware More...
AC input   240 V
feature   CE compliant
mfr. no.   Nedermans
availability   available only in EU
Description
General description
• Captures hazardous contaminants at source
• Extracts and filters up to 300m3/h
• Adjustable hood on 1.1m arm
• Two stage filter for vapours and particles
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?