• USA Home
  • Z214531 - ACE vacuum regulator
EMAIL THIS PAGE TO A FRIEND
Z214531 Aldrich
ACE vacuum regulator
Purchase
Description
General description
Improved cartesian type with increased sensitivity and versatility and reduced mercury requirement (20 to 30mL). A control point within 1mm Hg can be fine-tuned by adjusting the height of the pumpout tubulation. A spherical joint permits attitude adjustment to the orifice to accommodate a small continuous leak without trapping the float.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?