EMAIL THIS PAGE TO A FRIEND
Z214787 Aldrich
ACE SS turbine
size 75 mm
Purchase
Properties
Related Categories Ace, Ace Blades and Turbines, Ace SS Turbine, Ace Stirrer Blades, Ace Stirrers,
feature   fits shaft o.d. 10 mm
mfr. no.   Ace Glass 809531
size   75 mm
Description
General description
Multiple fins that may be bent to different angles. Set screw allows positioning at various locations on either glass or SS stirring shafts. Use SS set screw with SS stirring shaft. With glass shaft, use Kel-F tipped set screw. Both screws supplied with agitator.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?