EMAIL THIS PAGE TO A FRIEND
Z215414 Aldrich
Rigid PTFE sleeve
Joint: ST/NS 14/35
Synonym: ptfe sleeves
Purchase
Properties
Related Categories Cowie, Cowie Rigid PTFE sleeve, Cowie Stoppers, Joint Sleeves, Labware,
Joint   Joint: ST/NS14/35
mfr. no.   Cowie
Description
General description
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?