• USA Home
  • Z215627 - PTFE bushing-type reducing adapter
EMAIL THIS PAGE TO A FRIEND
Z215627 Aldrich
PTFE bushing-type reducing adapter
female Joint: ST/NS 34/45, male Joint: ST/NS 24/40
Purchase
Properties
Related Categories Bushing-type, Cowie, Cowie Adapters, Enlarging/Reducing Adapters, Glassware,
Joint   female Joint: ST/NS34/45
  male Joint: ST/NS24/40
mfr. no.   Cowie
Description
General description
PTFE with substantial grip-ring on shoulder. Grease-free, high-vacuum performance is obtained by transverse trapezoidal sealing rings on the contacting surfaces. Usable up to 280°C.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?