EMAIL THIS PAGE TO A FRIEND
Z217123 Aldrich
Thermo-Flasks
capacity 1.0 L
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Dewars, Barnstead/Thermolyne Dewars, Thermo-Flask Series, Cryoware, Labware,
material   stainless steel
sterility   non-sterile
feature   closure type Stainless Steel lids
  liq N2 retention 30 hr (At 80% capacity)
packaging   pack of 1
mfr. no.   Barnstead International 2122
I.D. × H   3.3 in. × 9 in.
capacity   1.0 L
Description
General description
Flasks have a borosilicate glass inner liner evacuated to 1 × 10-5 mm Hg for extended temperature retention of liquid nitrogen or solid CO2. Exteriors are corrosion-resistant SS with carrying handles and vented lids secured by two toggle clamps. Highly recommended for safe transport of liquid nitrogen and specimen storage.
Legal Information
Thermo-Flask is a trademark of Lab-Line Instruments, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?