• USA Home
  • Z217476 - Mini timer- 20 hours
EMAIL THIS PAGE TO A FRIEND
Z217476 Aldrich
Mini timer- 20 hours
Purchase
Properties
Related Categories Labware, Timers More...
Description
General description
6-digits, 1/2 in. H, counts up to 19 h, 59 min., 59 s in intervals of 1 min., warning tones at 10 and 5 min.; 20 s alarm when time is up. Includes 1.5 V button-cell battery.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
T6039 Quartz digital timers, Model T-7
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?