• USA Home
  • Z217506 - Indoor/outdoor thermometer
EMAIL THIS PAGE TO A FRIEND
Z217506 Aldrich
Indoor/outdoor thermometer
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
Description
General description
Similar to ambient temperature sensor above, but with an additional external sensor and 9ft cord. A switch changes the readout from indoor to outdoor temperature. Ideal for incubators or places where simultaneous internal/external readings are required.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z217492 Ambient temperature thermometer
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?