• USA Home
  • Z217565 - Flexible stirrer shaft coupling
EMAIL THIS PAGE TO A FRIEND
Z217565 Aldrich
Flexible stirrer shaft coupling
for glass shaft O.D., 19 mm, for motor shaft O.D., 1/2 in.
Purchase
Properties
Related Categories Flexible stirrer shafts and coupling, Glass Stirrer Shafts, Glass Stirring Equipment, Glassware, Labware More...
feature   for glass shaft O.D. 19 mm
  for motor shaft O.D. 1/2 in.
Description
General description
Flexible coupling allows angular and parallel misalignment of glass stirring shaft without breakage. Neoprene rubber sleeve body absorbs shock and vibration and has great torsional stiffness for quiet, smooth running. PTFE sleeve slides over end of glass shaft. SS set screw secures other end of coupling to motor shaft.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?