• USA Home
  • Z217743 - Michel-Miller adjustable bed height column
EMAIL THIS PAGE TO A FRIEND
Z217743 Aldrich
Michel-Miller adjustable bed height column
L × I.D. 300 mm × 25 mm
Purchase
Description
General description
Top fitting can be moved to compensate for changes in bed height from 1 to 6 in. below thread. Loosen bushing to slide end-fitting up and down. End-fitting is all PTFE with a glass disc, porosity 25 to 50 μm. Bottom of column is a #11 Ace-Thred bottom end-fitting. Top fitting has a connection for 2 mm I.D. PTFE tubing and bottom fitting has a 1/4 -28 tap for connecting small bore tubing.
Note: 10 mm column is same diameter top to bottom.
Price and Availability
All labs need water
Non-Sterile Millex Syringe Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replacement part
Product #
Description
Add to Cart
Z179639 Bottom end fitting for Michel-Miller adjustable bed height column
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?