• USA Home
  • Z218138 - Ace liquid-liquid extraction apparatus
EMAIL THIS PAGE TO A FRIEND
Z218138 Aldrich
Ace liquid-liquid extraction apparatus
Synonym: liquid liquid extraction, liquid liquid extractor, liquid-liquid extractor
Purchase
Properties
Related Categories Ace, Ace Extractors, Extractors, Labware, Liquid-Liquid Extraction,
mfr. no.   Ace Glass 684820
Description
General description
Designed for U.S. EPA "Priority Pollutant" analysis using Method 625 (base/neutral- and acid-extractables). Accommodates a 1 L sample and approximately 200 ml of a heavier-than-water extracting solvent. Flat extractor bottom permits use of a magnetic stirrer for efficient compound extraction from sample. Apparatus includes extractor body with one 24/40 joint clamp. PTFE stopper and 1 x 5, 2 mm bore PTFE stopcock for drainage, Allihn condenser, and a 1 L solvent flask.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?