• USA Home
  • Z218626 - Ace round-bottom reaction flask
EMAIL THIS PAGE TO A FRIEND
Z218626 Aldrich
Ace round-bottom reaction flask
capacity 1,000 mL
Purchase
Properties
Related Categories Ace, Ace Flasks, Ace Round-Bottom Reaction Flask, Labware, Manufacturer Browser More...
material   flask round bottom
  stainless steel
sterility   non-sterile
feature   closure type Lid
packaging   pack of 1
mfr. no.   Ace Glass 651106
capacity   1,000 mL
Description
General description
Cylindrical heavy-wall reaction flask with extra-thick flat flange. No constriction at top facilitates introduction or removal of material and allows easy cleaning. Order reaction heads and clamps separately below.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?