• USA Home
  • Z218677 - Ace reaction flask head w/ thermometer joint

EMAIL THIS PAGE TO A FRIEND
Z218677 Aldrich

Ace reaction flask head w/ thermometer joint

fits flask, 2,000 ‑ 4,000 mL, center Joint: ST/NS 29/42, side Joint: ST/NS 24/40

Synonym: flask lab, lab flask, reaction flasks

Purchase

Properties

Related Categories Ace, Ace Flasks, Ace Reaction Flask Heads, Labware, Manufacturer Browser More...
Joint   center Joint: ST/NS29/42
  side Joint: ST/NS24/40
feature   fits flask 2,000 ‑ 4,000 mL
mfr. no.   Ace Glass 651334

Description

General description

For use with reaction flasks above. Heads have three in-line joints and a #7 Ace-Thred, internally threaded side opening for thermometer (6 to 7 mm o.d.). Thread forms a compression seal with bushing and permits vertical adjustment of thermometer.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?