• USA Home
  • Z220396 - Aldrich® Schlenk-type adapter
EMAIL THIS PAGE TO A FRIEND
Z220396 Aldrich
Aldrich® Schlenk-type adapter
four-way adapter for solid-transfer vessel, male Joints: ST/NS 14/20 (3), female Joint: ST/NS 29/32
Purchase
Description
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?