• USA Home
  • Z221260 - Safe Storage of Laboratory Chemicals, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z221260 Aldrich
Safe Storage of Laboratory Chemicals, 2nd ed.
  •  ISBN-10 0471515817
  •  ISBN-13 978-047151581-4
Purchase
Properties
Related Categories Books and Software, Labware, Safety Books More...
publication info   D. Pipitone, ed., John Wiley & Sons, Ltd, 1991, hard cover
mfr. no.   Wiley
Description
General description
Divided into two parts, it begins with an introduction to fundamental principles of chemical storage and provides a logical framework for integrating storage concepts. The impact of federal legislation on chemical storage is discussed in detail. Storage requirements for hazardous, flammable, unstable, and incompatible chemicals are also covered. Further, it outlines procedures for labeling chemicals, addressing emergencies, and managing chemical safety with the aid of computers. In the second part, case studies cover such topics as surveys and inspections, large warehouse storage facilities, recommendations for industrial labs and a showcase university chemical storage facility.
Table of Contents
STORAGE GUIDELINES.
The Impact of Federal Regulations on the Storage of Laboratory Chemicals (D. Pipitone).
Storage Requirements for Flammable and Hazardous Chemicals (N. Steere).
Incompatible Chemicals in the Storeroom: Identification and Segregation (L. Bretherick).
Labeling Practices for Chemicals in Laboratories, Workplaces, and Chemical Storerooms (L. Nicholls).
Emergency Response for Releases of Laboratory Chemicals (D. Pipitone).
Computer Applications and Systems for Hazardous Materials and Waste Compliance in the Era of Regulation (A. Macenski).
CASE HISTORIES.
Chemical Storage for Industrial Laboratories (F. Chlad).
Implementation of an On-Line IMS Database System for Warehouse and Inventory Management (E. Houston).
Surveys and Inspections at Academic Chemical Storage Facilities (P. Redden).
University of Akron Chemical Storage Facilities (F. Chlad).
Safety First in the Elimination of Hazardous Chemicals from School Science Settings (J. Gerlovich).
Appendices.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?