• USA Home
  • Z221465 - Microscale Techniques For The Organic Laboratory
EMAIL THIS PAGE TO A FRIEND
Z221465 Aldrich
Microscale Techniques For The Organic Laboratory
  •  ISBN-10 0471621927
  •  ISBN-13 978-047162192-8
Purchase
Properties
publication info   D.W. Mayo, John Wiley & Sons, Ltd, 1991, 304 pp., soft cover
mfr. no.   Wiley
Description
General description
Covers the basic laboratory techniques of microscale organic chemistry. Beginning with an overview of lab safety, it goes on to cover equipment and techniques. Without touching on specific experiments, it includes a section on qualitative analysis. An introduction to and interpretation of IR theory and group frequencies is provided. Later sections cover NMR, 1H NMR spectra and 13C NMR spectroscopy. Also includes microscale amounts, which decrease the cost of chemicals, increase safety, improve air quality due to less waste disposal, and decrease the time needed to complete experiments.
Table of Contents
Safety.
Introduction to Microscale Laboratory Equipment and Technique.
Determination of Physical Properties.
Development of Microscale Techniques.
Introduction to Infrared Spectroscopy.
Introduction to Nuclear Magnetic Resonance Spectroscopy.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?