• USA Home
  • Z222062 - Aldrich® continuous extractor
EMAIL THIS PAGE TO A FRIEND
Z222062 Aldrich
Aldrich® continuous extractor
top Joint: ST/NS 24/40, bottom Joint: ST/NS 24/40
Purchase
Properties
Related Categories Aldrich, Aldrich Extractors, Continuous extractor, Extractors, Labware,
Joint   bottom Joint: ST/NS24/40
  top Joint: ST/NS24/40
plate I.D.   37 mm
Description
General description
Unique design eliminates fragile siphon tubes that can plug-up with recrystallized product. Top and bottom joints with a flat flange on the removable top. Separate inner glass tube has perforated bottom for support of filter paper. May also be used with standard paper or glass extraction thimbles.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?