• USA Home
  • Z222526 - Chem-FLEX tubing
EMAIL THIS PAGE TO A FRIEND
Z222526 Aldrich
Chem-FLEX tubing
Type 125, blue, I.D. 1/8 in., O.D. 0.4 in.
Purchase
Properties
Related Categories Aldrich, Aldrich Tubing, Chem-Flex Tubing, Labware, Manufacturer Browser,
I.D.   1/8 in.
O.D.   0.4 in.
color   blue
Description
General description
Developed by Aldrich for the safe transfer of aggressive solvents, air-sensitive liquids, and gases. Based on the original Aldrich Flex-needle concept, chemists can now fabricate their own transfer lines with CHEM-FLEX tubing to any length up to 50ft, easily and economically. CHEM-FLEX is suitable for many applications where standard PTFE tubing is used.
Available in four sizes to suit flow requirements. Order tubing, clamps, transfer needles and fittings separately below.
Request Technical Information Bulletin No. AL-192 for a complete listing of fittings and accessories.
Features:
• Chemically inert, thin-wall PTFE inner tube sheathed in clear PVC for extra strong, flexible transfer lines that are resistant to crushing or kinking
• Easily clamped to transfer needles and accessory fittings
• Color-coded tubing-size, imprinted with "safety stripe" for high visibility in the lab
• Cut-marks at 6 in. intervals
Legal Information
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?