• USA Home
  • Z223182 - Sound abating enclosure
EMAIL THIS PAGE TO A FRIEND
Z223182 Aldrich
Sound abating enclosure
For 130W model
Purchase
Properties
Related Categories Branson processors, Labware, Manufacturer Browser, Sonics & Materials, Sonics & Materials Sound Abating Enclosure,
mfr. no.   Sonics & Materials 630-0451
overall H × W × D   20 in. (51 cm) × 12 in. (30 cm) × 12 in. (30 cm)
Description
General description
Greatly reduces high-pitched noise produced by harmonics emanating from vessel walls and fluid surface during processing. Probe assembly is introduced through opening at top. Two access ports allow coolant or sample flow through the processing vessel while the door is closed. Waterproof, acoustically insulated interior and transparent access door.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?