• USA Home
  • Z223301 - Aldrich® high-dilution adapter
EMAIL THIS PAGE TO A FRIEND
Z223301 Aldrich
Aldrich® high-dilution adapter
Purchase
Description
General description
24/40 joints. Reactants added dropwise from an addition funnel are prediluted with approximately 25 mL of refluxing solvent held in a reservoir by an internal collar.1 Three top joints for connecting a reflux condenser, stirrer bearing/shaft (or stopper), and an addition funnel. Includes glass stopper.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?