EMAIL THIS PAGE TO A FRIEND
Z224162 Aldrich
Nylon tubing clamp
diam. 1/4- 5/16 in.
Purchase
Properties
Related Categories Chemistry Equipment and Labware, Clamps and Supports, Labware, Tubing
diam.   1/4- 5/16 in.
Description
General description
Non-corroding, lightweight clamps secure tubing to transfer needles, fittings, etc. Press clamp jaws together with pliers to secure.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?