• USA Home
  • Z224464 - Gilmont® direct reading flowmeter
EMAIL THIS PAGE TO A FRIEND
Z224464 Aldrich
Gilmont® direct reading flowmeter
tube size 14
Purchase
Properties
Related Categories Flowmeters, Gilmont, Gilmont Direct Reading Flowmeter, Labware, Manufacturer Browser,
tube size   14
mfr. no.   Gilmont EW-03232-24
measuring range   1,000-36,000 mL/min flow rate, (air, glass float)
  1,800-64,000 mL/min flow rate, (air, SS float)
  10-850 mL/min flow rate, (water, glass float)
  21-1,820 mL/min flow rate, (water, SS float)
parameter   ≤60 psi pressure
L × O.D.   9 3/4 in. × 1 1/2 in.
bushing NPTM   1/2 in.
Description
General description
For precise flow-monitoring of liquids and gases. Clear PC safety shield permits use at high temperatures and pressures. Fluid contacts PTFE body, borosilicate glass tube, Viton® O-rings, and floats (glass, 316 SS, or ruby).
NPT-male end bushings are PP with PTFE inserts that accept flexible hose. For high-purity applications, order PTFE coupling adapters to prevent fluids from contacting the bushings.
Accurate: ±5% of the reading or ±2 mm of the scale length (scale = 100 divisions). Take direct air and water readings without calibration curves. An "R factor" chart and formulas are included to convert scale readings for other fluids or for floats other than glass.
Accepts 3/8 in. I.D. tubing (sizes 14 and 15 accept 5/8 in. I.D. tubing).
Note: Flow rates are at standard temperature and pressure (20°C and 14.7 psi).
Legal Information
Gilmont is a registered trademark of Gilmont Instruments, Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
Z224529 PTFE capillary valve, Micrometer
Z224502 Pipe-to-tubing adapter, NPTM × O.D. 1/4 in. × 3/8 in.
Z224480 PTFE coupling adapter, NPTF 1/4 in.
Z224588 O-rings for PTFE metering capillary valve, PTFE
Z224553 Flowmeter stand kit, with built-in bubble level
Z224537 PTFE capillary valve, Metering
Z224510 Pipe-to-tubing adapter, NPTM × O.D. 1/2 in. × 1/2 in.
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?