• USA Home
  • Z224553 - Flowmeter stand kit
EMAIL THIS PAGE TO A FRIEND
Z224553 Aldrich
Flowmeter stand kit
with built-in bubble level
Purchase
Properties
Related Categories Flowmeter stand kit, Flowmeters, Gilmont, Labware, Manufacturer Browser,
mfr. no.   Gilmont
Description
General description
Includes aluminum base, rod, and 2 clamps to mount flowmeter on a stand or benchtop. Built-in bubble level for accuracy.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?