• USA Home
  • Z224766 - Aldrich® combustion apparatus
EMAIL THIS PAGE TO A FRIEND
Z224766 Aldrich
Aldrich® combustion apparatus
Apparatus
Purchase
Properties
Related Categories Aldrich, Aldrich Flasks, Aldrich Speciality Flasks, Flasks, Labware,
material   borosilicate glass
  flask flat bottom
sterility   non-sterile
packaging   pack of 1
mfr. no.   Sigma-Aldrich
capacity   1000 mL
Description
General description
1,000 mL heavy-wall borosilicate glass conical flask with 35/25 ball joint stopper. Detachable platinum sample carrier is suspended from stopper hook. Pinch clamp holds stopper in place during combustion. Includes flask, stopper, platinum carrier, and pinch clamp. Order sample wrappers separately.
Based on a design by C.L. Ogg, R.B. Kelly, and J.A. Connelly. See Microchemical Techniques, N. D. Cheronis, Ed., Interscience, 1962, pp 427-433.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replacement part
Product #
Description
Add to Cart
Z224871 Pinch clamps for SJ joints, Replacement pinch clamp for Aldrich combustion apparatus
Z224855 Replacement flask for Aldrich® combustion apparatus, capacity 1000 mL
Z224898 Aldrich® combustion apparatus, Replacement sample carrier, Pt
Z224863 Replacement stopper for Aldrich® combustion apparatus, Replacement stopper
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?