• USA Home
  • Z224774 - Aldrich® condenser for Büchi® rotary evaporators
EMAIL THIS PAGE TO A FRIEND
Z224774 Aldrich
Aldrich® condenser for Büchi® rotary evaporators
Purchase
Properties
Related Categories Dry-Ice (Buchi) High-Capacity Condensers, Evaporators, Glass Condensers, Glassware, High-Capacity Condensers,
Joint   female Joint ST/NS19/38
  male Joint SJ35/25
description   No. 50 flange
H   370 mm
Description
General description
For Büchi R110, R114, and R124 evaporators.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Büchi is a registered trademark of Büchi Labortechnik AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?