• USA Home
  • Z224820 - Calibra® digital micropipette
EMAIL THIS PAGE TO A FRIEND
Z224820 Aldrich
Calibra® digital micropipette
100-1095 μL volume, precision: 0.1%
Purchase
Properties
mfr. no.   Wheaton Scientific
measuring range   100-1095 μL volume, precision: 0.1%
color   blue Fits microtips
Description
General description
Positive displacement and solid calibration provides absolute accuracy and reproducibility. Variable volume pipette utilizes a cylindrical cam assembly with permanently fixed calibrated stops, each one corresponding to one volume setting. Does not lose original calibration, even after prolonged use. Order micropipette tips below.
Legal Information
Calibra is a registered trademark of Socorex Isba S.A.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?