• USA Home
  • Z225797 - Aldrich® cold-finger for high-efficiency cold-trap
EMAIL THIS PAGE TO A FRIEND
Z225797 Aldrich
Aldrich® cold-finger for high-efficiency cold-trap
Silvered and evacuated, Joint: ST/NS 45/50
Purchase
Description
General description
Extremely efficient design minimizes warming of liquid nitrogen or dry ice/acetone coolant that is added through a side port in the wall of the Dewar and then plugged with a vented stopper. Order Dewar, Z225770, separately.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?