• USA Home
  • Z228109 - 3M dust, mist and fume respirator
EMAIL THIS PAGE TO A FRIEND
Z228109 Aldrich
3M dust, mist and fume respirator
size M/L
Purchase
Properties
Related Categories 3M, Disposable Respirators, Dust, mist and fume respirator, Labware, Manufacturer Browser,
size   M/L
feature   Class FFP3SL
mfr. no.   3M 8835
availability   available only in EU
Description
General description
This respirator is exceptionally comfortable with a soft face piece sealing ring, low breathing resistance, and an exhalation valve to reduce moisture build-up. Protects against fine toxic dusts, fibres, mists and fume. Maximum usage level of 50 × OEL for fine toxic dusts and mists, 25 × OEL for fine toxic fumes.
Legal Information
3M is a trademark of 3M Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?