• USA Home
  • Z228273 - Bollé Cover all chemical goggles
EMAIL THIS PAGE TO A FRIEND
Z228273 Aldrich
Bollé Cover all chemical goggles
spare lens
Purchase
Properties
Related Categories Eye Protection Equipment, Goggles, Labware, Safety Equipment
mfr. no.   Bollé
availability   not available in USA and Canada
Description
General description
General use goggles with a clear polycarbonate lens that provides grade 1 impact protection as well as meets the British Standards for chemical splash, dusts and molten metals. BS2092.1CDM
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?