| Related Categories | Bolle, Bolle Goggles, Eye Protection Equipment, Goggles, Labware, |
| material | glass (carbo) |
| availability | not available in USA and Canada |
These googles have a soft, lightweight PVC frame and a foam surround which provide good visibility and a tight comfortable fit for protection against chemical splash, dust and gases. They are supplied with a no fog len for greater chemical resistance.