• USA Home
  • Z228389 - Clearways faceshield
EMAIL THIS PAGE TO A FRIEND
Z228389 Aldrich
Clearways faceshield
Acetate visor, 200 mm
Purchase
Properties
Related Categories Eye Protection Equipment, Faceshields, Labware, Safety Equipment
mfr. no.   Pulsafe
availability   not available in USA and Canada
Description
General description
A lightweight browguard with elastic headband and comfort band combined with a 200 mm acetate visor or a 200 mm metal bound polycarbonate visor to provide the wearer with a choice of protection.
• Acetate visor: BS2092.2C
• Polycarbonate (PC) visor: BS2092.1C
Order browguard and desired visor separately.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z228397 Clearways faceshield, Polycarbonate visor only, 200 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?