Z228397 Aldrich
Clearways faceshield
Polycarbonate visor, 200 mm
DOWNLOAD MSDS (PDF)
Properties
Related Categories Eye Protection Equipment, Faceshields, Labware, Safety Equipment
availability   not available in USA and Canada
Description
General description
A lightweight browguard with elastic headband and comfort band combined with a 200 mm acetate visor or a 200 mm metal bound polycarbonate visor to provide the wearer with a choice of protection.
• Acetate visor: BS2092.2C
• Polycarbonate (PC) visor: BS2092.1C
Order browguard and desired visor separately.
Price and Availability
Customers Also Viewed
Clearways faceshield
Browguard
Clearways faceshield
Acetate visor, 200 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?