• USA Home
  • Z229725 - Thermolyne® 21100 tube furnace
EMAIL THIS PAGE TO A FRIEND
Z229725 Aldrich
Thermolyne® 21100 tube furnace
AC input 120 V
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Ovens, Heating, Labware, Manufacturer Browser,
feature   CE compliant
  CSA approved
mfr. no.   Barnstead International F21135
W × H × D   40.7 cm × 37.4 cm × 31.0 cm
output   11.2 A
power consumption   1250 W
Description
General description
Features accurate, automatic temperature control with self-tuning function that maintains preset temperature. LCD display shows both set point and chamber temperature (100 to 1,200 °C). Heating elements embedded in ceramic fiber insulation for rapid heatup and cool down. Adjustable alarm or OTP temperature setting can be used to protect the furnace or loads in the chamber from excess heat. Chamber supplied with two interchangeable end pieces to accept tube diameters 2.5 and 5.0 cm. Furnace operates in any position. Includes line cord and plug.
Legal Information
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?