• USA Home
  • Z230340 - Best® N-DEX® laboratory gloves
EMAIL THIS PAGE TO A FRIEND
Z230340 Aldrich
Best® N-DEX® laboratory gloves
lightly powdered, cornstarch, thickness 4 mil (0.12 mm), size 9‑10 XL
Purchase
Properties
Related Categories Best Gloves, Best N-DEX Laboratory Gloves, Disposable Gloves, Disposable Nitrile Gloves, Gloves,
material   nitrile
size   9‑10 XL
sterility   non-sterile
feature   disposable
  latex free: No
  lightly powdered cornstarch
packaging   pkg of 100 ea
mfr. no.   N-DEX® 7005
L   240 mm
thickness   4 mil (0.12 mm)
color   blue
Description
General description
Soft elastic material feels cooler than natural rubber or latex and conforms to hand for greater comfort and dexterity.
• 100% nitrile non-latex glove
• Ambidextrous
• Excellent alternative to latex
• Rolled cuff
Legal Information
Best is a registered trademark of Showa Glove Co.
N-DEX is a registered trademark of Showa Glove Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?