EMAIL THIS PAGE TO A FRIEND
Z230642 Aldrich
Oberon lenses
didymium II blue
Purchase
Properties
Related Categories Eye Protection Accessories, Eye Protection Equipment, Labware, Safety Equipment
material   polycarbonate lens
availability   not available in EU
frame size   58 mm
color   didymium II blue
  tan Frame
Description
General description
Lenses are coated with both a gold coating (which will not wrinkle or warp) providing UV and IR protection plus Silicote 1 for scratch resistance.
Wire core adjustable temples and metal hinges. Meets ANSI Z87.1-1989 requirements.
Removes yellows while maintaining reds and oranges to enable visibility while heating glass.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?