• USA Home
  • Z231681 - Duran® crystallizing dishes
EMAIL THIS PAGE TO A FRIEND
Z231681 Aldrich
Duran® crystallizing dishes
capacity 60 mL
Purchase
Properties
Related Categories Dishes; Life-Science, Duran, Duran Dishes, Glass Crystallizing Dishes, Glass Dishes,
mfr. no.   DURAN
  Schott
availability   not available in USA and Canada
H × O.D.   35 mm × 60 mm
capacity   60 mL
Description
General description
Cylindrical dishes without spout. Manufactured from Duran borosilicate glass type 3.3 for maximum chemical and thermal shock resistance. Meets DIN 12337 standard.
Legal Information
Duran is a registered trademark of Duran Group
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?