• USA Home
  • Z232955 - Duran® Erlenmeyer flasks with screw cap
EMAIL THIS PAGE TO A FRIEND
Z232955 Aldrich
Duran® Erlenmeyer flasks with screw cap
capacity 500 mL
Synonym: conical flasks
Purchase
Properties
Related Categories Erlenmeyer Flasks, Glass, Flasks, Labware More...
material   borosilicate glass
  narrow-mouth
neck thread size   GL 32
sterility   non-sterile
feature   closure type Screw cap
packaging   pack of 10
mfr. no.   DURAN
  Schott
availability   not available in ROW (Available only in EU countries)
  not available in USA and Canada
H × O.D.   180 mm × 105 mm
capacity   500 mL
Description
General description
Graduated and complete with screw cap. Caps will withstand autoclaving temperatures. Manufactured from Duran borosilicate glass 3.3 for maximum chemical and thermal shock resistance.
Legal Information
Duran is a registered trademark of Duran Group
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?