• USA Home
  • Z233374 - High-Performance Liquid Chromatography of Peptides and Proteins: Separation, Analysis, and Conformation
EMAIL THIS PAGE TO A FRIEND
Z233374 Aldrich
High-Performance Liquid Chromatography of Peptides and Proteins: Separation, Analysis, and Conformation
  •  ISBN-10 084936549X
  •  ISBN-13 978-084936549-2
Purchase
Properties
publication info   C.T. Mant, CRC Press, Boca Raton, FL, 1991, 960 pp., hard cover
mfr. no.   CRC Press
Description
General description
This book consists of a series of 82 precise, easy-to-read articles by internationally renowned scientists and emphasizes the practical approach to HPLC with minimal theory, although the underlying principles to peptide and protein separations are clearly expressed. All of the major modes of microbore, ultrafast and analytical HPLC are discussed, including size-exclusion, ion-exchange, reversed-phase, hydrophobic interaction, and affinity and immunoaffinity chromatography. A section on preparative HPLC, including displacement techniques, is also presented. Problem-solving approaches to the
separation of various classes of biologically active peptides and proteins are thoroughly explored, while the importance of peptide standards for monitoring column performance and for optimizing separation conditions is emphasized.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?