• USA Home
  • Z234109 - Kilo-Lab® cylinder
EMAIL THIS PAGE TO A FRIEND
Z234109 Aldrich
Kilo-Lab® cylinder
capacity 90 L , nominal
Purchase
Description
General description
For storing and dispensing laboratory-scale to production quantities of air-sensitive liquids.
Technical Information Bulletin No. AL-149, with instructions for use of the system and a list of fittings and accessories, is included with each cylinder. Meets DOT 4BA-240, rated 240 psi.
• Cylinder does not come with valves won′t have valves
• Cylinder is not TC stamped as required in Canada
• The 90 L cylinder is pictured
With protective cover. Mild steel, without valves.
Legal Information
Kilo-Lab is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?