• USA Home
  • Z235830 - Infrared Absorbing Dyes
EMAIL THIS PAGE TO A FRIEND
Z235830 Aldrich
Infrared Absorbing Dyes
  •  ISBN-10 0306434784
  •  ISBN-13 978-030643478-5
Purchase
Properties
publication info   M. Matsuoka, ed., Kluwer Academic Publishers, 1990, 234 pp., hard cover
mfr. no.   Kluwer Academic Publishers
Description
General description
Reviews the synthesis and characteristics of infrared dyes, which can now be designed on personal computers, and their application in high technology, especially their critical importance in the expanding use of laser diodes as a light source. The 15 separately authored chapters describe the synthesis of dye groups such as cyanine, quinone, metal complex, and photochronic; and applications in semiconductors, optical recording, printing, medicine, and other fields
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?