• USA Home
  • Z235938 - Polymer Syntheses, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z235938 Aldrich
Polymer Syntheses, 2nd ed.
Volume 1
  •  ISBN-10 0126185115
  •  ISBN-13 978-012618511-9
Purchase
Properties
Related Categories Books and Software, Labware, Polymer Chemistry Books More...
publication info   S. Sandler and W. Karo, Academic Press, 1991, 602 pp., hard cover
mfr. no.   Elsevier
Description
General description
Provides detailed laboratory instructions for the synthesis of polymers.
Table of Contents
Polymerization of olefinic and diolefinic hydrocarbons
Polyesters
Polycarbonates
Polyamides
Polymerization of aldehydes
Polymerization of epoxides and cyclic ethers
Polyureas
Polyurathanes
Thermally stable polymers
Polymerization of acrylate and methacrylate esthers
Polymerization of nitrile monomers
Polyacrylamide and related amides
Organophosphorus polymers
Free-radical initiation of vinyl and related monomers
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?