• USA Home
  • Z236667 - Willson® Orion® safety glasses

EMAIL THIS PAGE TO A FRIEND
Z236667 Aldrich

Willson® Orion® safety glasses

lens size 54 mm, black Frame

Purchase

Properties

Related Categories Eye Protection Equipment, Eyeshields, Labware, Manufacturer Browser, Safety Equipment,
mfr. no.   Willson
availability   not available in EU
  not available in EU
lens size   54 mm
color   black Frame
  clear Lens

Description

General description

54mm frame size

Designed to fit the most demanding worker, this rugged, lightweight eyewear provides all-day comfort and protection. Aviator-style metal frame with one-piece nosepad. Meets ANSI Z87.1-1989 and CSA-Z94.3-M92 requirements.

Legal Information

Orion is a registered trademark of Thermo Electron Corp.

Willson is a registered trademark of Willson Safety Products

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?