EMAIL THIS PAGE TO A FRIEND
Z236926 Aldrich
Gas-cylinder cart
floorstand, capacity, 4 cylinders
Purchase
Properties
Related Categories Gas Equipment Product Guide, Gas Storage, Labware More...
feature   capacity 4 cylinders
H × W × D   30 in. × 36 1/2 in. × 24 in.
weight   92 lb
Description
General description
Durable supports hold gas, oxygen, welding, cascade, propane, or LP cylinders, with diameters 3 to 123/8 in. Heavy 11 gauge welded construction, baked enamel finish, steel-reinforced resin edge-guards. 11/2 in. PP webbed straps with steel cinch buckles and chain sets assure long service life. All units comply with OSHA, NFPA, UFC, and AGA standards.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?