Z238147 Aldrich
Aldrich® LB-cylinder holder
Bench top, capacity, 1 cylinder
DOWNLOAD MSDS (PDF)
Properties
Related Categories Aldrich, Aldrich Gas Equipment, Aldrich Supports (Gas), Gas Cylinders and Accessories, Gas Equipment Product Guide,
material   PVC
feature   capacity 1 cylinder
W × H × D   8 in. × 5 in. × 8 in.
Description
General description
Hold up to 2 in. diameter cylinders. Bench/wall mountable units have four countersunk screw holes for attachment.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Customers Also Viewed
Aldrich<SUP>®</SUP> LB-cylinder holder
Bench/wall mount, capacity, 1 cylinder
Aldrich<SUP>®</SUP> LB-cylinder holder
Bench/wall mount, capacity, 3 cylinders
Aldrich<SUP>®</SUP> LB-cylinder holder/porter
W × H × D 8 3/4 in. × 18 3/4 in. × 5 3/4 in., PVC
Lecture bottle
capacity 440 mL, 3/8 in. NPTF valve thread
Lecture-bottle control valve
Monel®, hose outlet I.D. 1/4 in.
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?