• USA Home
  • Z238163 - Aldrich® LB-cylinder holder
EMAIL THIS PAGE TO A FRIEND
Z238163 Aldrich
Aldrich® LB-cylinder holder
Bench/wall mount, capacity, 3 cylinders
Purchase
Properties
Related Categories Aldrich, Aldrich Gas Equipment, Aldrich Supports (Gas), Gas Cylinders and Accessories, Gas Equipment Product Guide,
material   PVC
feature   capacity 3 cylinders
W × H × D   11 in. × 7 3/4 in. × 4 in.
Description
General description
Hold up to 2 in. diameter cylinders. Bench/wall mountable units have four countersunk screw holes for attachment.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?