• USA Home
  • Z238171 - Aldrich® LB-cylinder holder/porter
EMAIL THIS PAGE TO A FRIEND
Z238171 Aldrich
Aldrich® LB-cylinder holder/porter
W × H × D 8 3/4 in. × 18 3/4 in. × 5 3/4 in., PVC
Purchase
Properties
Related Categories Aldrich, Aldrich Gas Equipment, Aldrich Supports (Gas), Gas Cylinders and Accessories, Gas Equipment Product Guide,
material   PVC
W × H × D   8 3/4 in. × 18 3/4 in. × 5 3/4 in.
Description
General description
Allows for up to six LB gas cylinders to be moved or stored at one time.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?