• USA Home
  • Z238791 - Introduction to Physical Polymer Science, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z238791 Aldrich
Introduction to Physical Polymer Science, 2nd ed.
  •  ISBN-10 0471530352
  •  ISBN-13 978-047153035-0
Purchase
Properties
publication info   L. Sperling, John Wiley & Sons, Ltd, 1992, hard cover
mfr. no.   Wiley
Description
General description
Updated and revised, it focuses on the role of molecular conformation and configuration in determining the physical behavior of polymers. New features include the amorphous and crystalline states of polymers; macromolecular hypothesis and historical development of photophysics and fluorescence; thermodynamics of blending polymers and polymer/polymer phase diagrams; a discussion of rheology plus gelatinous materials; and a variety of contemporary topics emphasizing surface, interfacial and electrical behavior of polymers, nonlinear optics and high temperature substances. Each chapter includes several classroom demonstrations and problem sets.
Table of Contents
Chain Structure and Configuration.
Molecular Weights and Sizes.
Concentrated Solutions and Phase Separation Behavior.
The Amorphous State.
The Crystalline State.
Polymers in the Liquid Crystalline State.
Glass-Rubber Transition Behavior.
Cross-Linked Polymers and Rubber Elasticity.
Polymer Viscoelasticity and Rheology.
Mechanical Behavior of Polymers.
Modern Topics.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?