• USA Home
  • Z238813 - Homogeneous Catalysis, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z238813 Aldrich
Homogeneous Catalysis, 2nd ed.
  •  ISBN-10 0471538299
  •  ISBN-13 978-047153829-5
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   G. Parshall, John Wiley & Sons, Ltd, 1992, hard cover
mfr. no.   Wiley
Description
General description
Contains a balanced discussion of homogeneous catalytic reactions that are used in industry, featuring every documented example employed in a current commercial process, or that have a broad application in the organic synthesis laboratory. Incorporates synthesis with chiral catalysts in chapters on hydrogenation, CO chemistry and olefin oxidation. New additions include Tennessee Eastman′s coal-based acetic anhydride plant and IFP′s Dimersol process for dimerizing propylene as well as major changes in the areas on pharmaceuticals, flavors, fragrances, agricultural and electronic chemicals.
Table of Contents
Trends in Homogeneous Catalysis in Industry.
Isomerization of Olefins.
Reactions of Olefins and Dienes--Hydrogenation and HY Additions.
Polymerization and Oligomerization of Olefins and Dienes.
Reactions of Carbon Monoxide.
Oxidation of Olefins and Dienes.
Arene Reactions.
Reactions of Acetylenes.
Carbene Complexes in Olefin Metathesis and Ring-Forming Reactions.
Oxidation of Hydrocarbons by Oxygen.
Esterification, Polycondensation, and Related Processes.
Homogeneous Catalysis in Halocarbon Chemistry.
Appendix.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?