• USA Home
  • Z239097 - Humidity/temperature monitor
EMAIL THIS PAGE TO A FRIEND
Z239097 Aldrich
Humidity/temperature monitor
23-122 °F temperature, accuracy: 2 °F, resolution: 1 °F, 20-99 %RH humidity, accuracy: 6%RH, resolution: 1%RH
Purchase
Properties
Related Categories Electronic Meters, Labware, Temperature More...
measuring range   -10-50 °C temperature, accuracy: 1 °C, resolution: 0.1 °C
  20-99 %RH humidity, accuracy: 6%RH, resolution: 1%RH
  23-122 °F temperature, accuracy: 2 °F, resolution: 1 °F
size   4 1/4 in. × 3 3/4 in. × 7/8 in.
Description
General description
Monitors both temperature and humidity simultaneously on a large 33/16 × 21/2 in. digital display. For desiccators, incubators, refrigerators, fume hoods, storage areas, etc. Recalls Min/Max temperature and humidity readings over any time period. Stand or wall mount. Includes 1 AAA battery.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?