• USA Home
  • Z242616 - Whatman® disposable filter funnels
EMAIL THIS PAGE TO A FRIEND
Z242616 Aldrich
Whatman® disposable filter funnels
(3, Qualitative)
Purchase
Properties
material   polypropylene (medical grade)
mfr. no.   Whatman
Description
Features and Benefits
Features and benefits:
• Disposable design eliminates dedicated glassware, ideal for radioactive applications
• Chemically resistant polypropylene housing allows for use with a wide range of aggressive solutions
• Designed for single use or batch sample processing
• Robotic friendly
• Removable filter allows for further processing
• Luer taper outlet for easy vacuum attachment
General description
The Whatman Disposable Filter Funnels, available in 25 and 47 mm diameters, are convenient filter funnels containing Whatman brand filter media. The filter media can be easily removed for further analysis. The unit is composed of medical grade polypropylene, compatible with most solutions.
25 mm Disposable Filter Funnel
The 25 mm Disposable Filter Funnel is a convenient, disposable filter funnel containing Whatman brand filter media. This 25 mm diameter filter can be used in evaluation of processed proteins in TCA precipitation or binding assay procedures and can be easily removed for further analysis or culturing.
The 25 mm Disposable Filter Funnel is available with glass microfiber filters. Typical applications include TCA precipitation, cell harvesting, tissue washing, protein precipitation, and high recovery capture filtration.
47 mm Disposable Filter Funnel
The 47 mm Disposable Filter Funnel is a convenient, disposable unit containing Whatman brand media. The 47 mm diameter filter can be easily removed for further analysis or culturing.
Features and benefits:
• 47 mm diameter Whatman brand filter
• Retrievable filter for further analysis
• Disposable for cleanliness and convenience
• 250 mL reservoir
• 0.45 μm cellulose nitrate available sterile for culturing
Chemical Resistance
Both 25 and 47 mm diameter filter funnels are compatible with aqueous solutions and most organic solvents. Caution should be used when working with strong acids or strong bases in these filter funnels.
Legal Information
Whatman is a registered trademark of GE Healthcare
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?